3-Amino-1-[(2,6-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR32956-1G
APOSOR32956-1GEA 1570 EUR
APOSOR32956-1G
3-Amino-1-[(2,6-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one, tech
3-Amino-1-[(2,6-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one
Formel: C₁₂H₁₀Cl₂N₂O
Molecular Weight: 269,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140253
CAS-Nummer: 338783-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR