Methyl (2E)-2-[(3-chlorophenyl)formamido]-3-(dimethylamino)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32957-1G
APOSOR32957-1GEA 1570 EUR
APOSOR32957-1G
Methyl (2E)-2-[(3-chlorophenyl)formamido]-3-(dimethylamino)prop-2-enoate, tech
Methyl (2E)-2-[(3-chlorophenyl)formamido]-3-(dimethylamino)prop-2-enoate
Formel: C₁₃H₁₅ClN₂O₃
Molecular Weight: 282,723 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00232790
CAS-Nummer: 400084-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR