N'-[(1E)-(Dimethylamino)methylidene]benzohydrazide, tech

Lieferant: Apollo Scientific

OR32961-1G
APOSOR32961-1GEA 1570 EUR
APOSOR32961-1G
N'-[(1E)-(Dimethylamino)methylidene]benzohydrazide, tech
N'-[(1E)-(Dimethylamino)methylidene]benzohydrazide
Formel: C₁₀H₁₃N₃O
Molecular Weight: 191,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140517
CAS-Nummer: 308239-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR