N'-[(1E)-(Dimethylamino)methylidene]pyridine-4-carbohydrazide, tech
Lieferant: Apollo Scientific
OR32962-1G
APOSOR32962-1GEA
1570
EUR
APOSOR32962-1G
N'-[(1E)-(Dimethylamino)methylidene]pyridine-4-carbohydrazide, tech
N'-[(1E)-(Dimethylamino)methylidene]pyridine-4-carbohydrazide
Formel:
C₉H₁₂N₄O Molecular Weight: 192,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00140519 CAS-Nummer: 16825-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...