4-Chloro-N'-[(1E)-(dimethylamino)methylidene]benzohydrazide, tech

Lieferant: Apollo Scientific

OR32963-1G
APOSOR32963-1GEA 1570 EUR
APOSOR32963-1G
4-Chloro-N'-[(1E)-(dimethylamino)methylidene]benzohydrazide, tech
4-Chloro-N'-[(1E)-(dimethylamino)methylidene]benzohydrazide
Formel: C₁₀H₁₂ClN₃O
Molecular Weight: 225,675 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140520
CAS-Nummer: 308239-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR