4-Brom-1-phenyl-1H-pyrazol 97%

Lieferant: Apollo Scientific
Achtung

OR3297-1G
APOSOR3297-1GEA 21.9 EUR
APOSOR3297-1G
4-Brom-1-phenyl-1H-pyrazol 97%
4-Bromo-1-phenyl-1H-pyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234505
CAS-Nummer: 15115-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR