2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile, tech

Lieferant: Apollo Scientific

OR32972-1G
APOSOR32972-1GEA 1570 EUR
APOSOR32972-1G
2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile, tech
2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile
Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231856
CAS-Nummer: 53813-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR