Methyl 1-benzyl-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32973-1G
APOSOR32973-1GEA 1570 EUR
APOSOR32973-1G
Methyl 1-benzyl-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl 1-benzyl-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794601
CAS-Nummer: 77867-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR