3-[4-(3-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile, tech

Lieferant: Apollo Scientific

OR32976-1G
APOSOR32976-1GEA 0 EUR
APOSOR32976-1G
3-[4-(3-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile, tech
3-[4-(3-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile
Formel: C₁₃H₁₅ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00664792
CAS-Nummer: 338794-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR