3-Amino-3-(4-methylpiperazino)acrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: (2E)-3-Amino-3-(4-methylpiperazin-1-yl)prop-2-enenitrile

OR32977-1G
APOSOR32977-1GEA 671 EUR
APOSOR32977-1G
3-Amino-3-(4-methylpiperazino)acrylonitrile 95%
3-Amino-3-(4-methylpiperazino)acrylonitrile
Formel: C₈H₁₄N₄
Molecular Weight: 166,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664794
CAS-Nummer: 136062-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR