3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile 95%

Lieferant: Apollo Scientific

Synonyme: (2Z)-3-Amino-3-[4-(3,4-dichlorophenyl)piperazin-1-yl]prop-2-enenitrile

OR32978-1G
APOSOR32978-1GEA 671 EUR
APOSOR32978-1G
3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile 95%
3-Amino-3-[4-(3,4-dichlorophenyl)piperazino]acrylonitrile
Formel: C₁₃H₁₄Cl₂N₄
Molecular Weight: 297,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664795
CAS-Nummer: 1164541-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR