Ethyl 4-hydroxy-6,8-dimethylquinoline-3-carboxylate 95%

Lieferant: Apollo Scientific

OR32979-1G
APOSOR32979-1GEA 671 EUR
APOSOR32979-1G
Ethyl 4-hydroxy-6,8-dimethylquinoline-3-carboxylate 95%
Ethyl 4-hydroxy-6,8-dimethylquinoline-3-carboxylate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173389
CAS-Nummer: 77156-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR