Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate 95%

Lieferant: Apollo Scientific

OR32982-1G
APOSOR32982-1GEA 224 EUR
APOSOR32982-1G
Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate 95%
Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
Formel: C₁₃H₁₀ClNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00173405
CAS-Nummer: 26893-17-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR