Methyl 2-[(2,5-dichlorophenyl)formamido]acetate, tech

Lieferant: Apollo Scientific

OR32988-1G
APOSOR32988-1GEA 1570 EUR
APOSOR32988-1G
Methyl 2-[(2,5-dichlorophenyl)formamido]acetate, tech
Methyl 2-[(2,5-dichlorophenyl)formamido]acetate
Formel: C₁₀H₉Cl₂NO₃
Molecular Weight: 262,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01195627
CAS-Nummer: 338965-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR