3-Chloro-N-(4-methoxyphenyl)-2,2-dimethylpropanamide, tech

Lieferant: Apollo Scientific

OR32992-1G
APOSOR32992-1GEA 1570 EUR
APOSOR32992-1G
3-Chloro-N-(4-methoxyphenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(4-methoxyphenyl)-2,2-dimethylpropanamide
Formel: C₁₂H₁₆ClNO₂
Molecular Weight: 241.714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793409
CAS-Nummer: 338968-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR