(3E)-4-[(4-Methoxyphenyl)amino]-3-phenylbut-3-en-2-one, tech

Lieferant: Apollo Scientific

OR32996-1G
APOSOR32996-1GEA 1570 EUR
APOSOR32996-1G
(3E)-4-[(4-Methoxyphenyl)amino]-3-phenylbut-3-en-2-one, tech
(3E)-4-[(4-Methoxyphenyl)amino]-3-phenylbut-3-en-2-one
Formel: C₁₇H₁₇NO₂
Molecular Weight: 267,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975146
CAS-Nummer: 1164560-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR