(3E)-4-[(4-Acetylphenyl)amino]-3-phenylbut-3-en-2-one, tech

Lieferant: Apollo Scientific

OR32997-1G
APOSOR32997-1GEA 1570 EUR
APOSOR32997-1G
(3E)-4-[(4-Acetylphenyl)amino]-3-phenylbut-3-en-2-one, tech
(3E)-4-[(4-Acetylphenyl)amino]-3-phenylbut-3-en-2-one
Formel: C₁₈H₁₇NO₂
Molecular Weight: 279,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975147
CAS-Nummer: 343373-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR