Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate, tech

Lieferant: Apollo Scientific

OR33000-1G
APOSOR33000-1GEA 1570 EUR
APOSOR33000-1G
Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate, tech
Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate
Formel: C₁₂H₁₇NO₃
Molecular Weight: 223,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316103
CAS-Nummer: 343373-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR