[5-(2-Chlorophenyl)-1,2-oxazol-3-yl]methanol, tech

Lieferant: Apollo Scientific

OR33004-1G
APOSOR33004-1GEA 1570 EUR
APOSOR33004-1G
[5-(2-Chlorophenyl)-1,2-oxazol-3-yl]methanol, tech
[5-(2-Chlorophenyl)-1,2-oxazol-3-yl]methanol
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01444102
CAS-Nummer: 343374-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR