5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one, tech
Lieferant: Apollo Scientific
OR33006-1G
APOSOR33006-1GEA
1570
EUR
APOSOR33006-1G
5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one, tech
5-Amino-6-methyl-4H,5H-furo[3,2-c]pyridin-4-one
Formel:
C₈H₈N₂O₂ Molecular Weight: 164,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01443847 CAS-Nummer: 57053-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...