3-[(Pyridin-3-yl)methoxy]benzoic acid hydrochloride 97%

Lieferant: Apollo Scientific

OR33008-1G
APOSOR33008-1GEA 1070 EUR
APOSOR33008-1G
3-[(Pyridin-3-yl)methoxy]benzoic acid hydrochloride 97%
3-[(Pyridin-3-yl)methoxy]benzoic acid hydrochloride
Formel: C₁₃H₁₁NO₃·ClH
Molecular Weight: 265,692 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28016109
CAS-Nummer: 1610377-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR