Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33009-1G
APOSOR33009-1GEA 1570 EUR
APOSOR33009-1G
Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate, tech
Methyl 1-(3-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylate
Formel: C₁₂H₉ClN₂O₃
Molecular Weight: 264,664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871606
CAS-Nummer: 478063-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR