N'-(6-Chloropyridin-2-yl)propanehydrazide, tech
Lieferant: Apollo Scientific
OR33012-1G
APOSOR33012-1GEA
1570
EUR
APOSOR33012-1G
N'-(6-Chloropyridin-2-yl)propanehydrazide, tech
N'-(6-Chloropyridin-2-yl)propanehydrazide
Formel:
C₈H₁₀ClN₃O Molecular Weight: 199,638 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02187189 CAS-Nummer: 66999-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...