1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)propan-1-ol, tech
Lieferant: Apollo Scientific
OR33014-1G
APOSOR33014-1GEA
1570
EUR
APOSOR33014-1G
1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)propan-1-ol, tech
1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)propan-1-ol
Formel:
C₉H₁₃Cl₂NOS Molecular Weight: 254,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02661707 CAS-Nummer: 439110-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...