(2E)-1-(Pyridin-2-yl)-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33016-1G
APOSOR33016-1GEA 1570 EUR
APOSOR33016-1G
(2E)-1-(Pyridin-2-yl)-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)prop-2-en-1-one, tech
(2E)-1-(Pyridin-2-yl)-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)prop-2-en-1-one
Formel: C₁₇H₁₆N₂O
Molecular Weight: 264,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187205
CAS-Nummer: 1823194-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR