2-Chloro-N'-(6-chloropyridin-2-yl)acetohydrazide, tech
Lieferant: Apollo Scientific
OR33017-1G
APOSOR33017-1GEA
1570
EUR
APOSOR33017-1G
2-Chloro-N'-(6-chloropyridin-2-yl)acetohydrazide, tech
2-Chloro-N'-(6-chloropyridin-2-yl)acetohydrazide
Formel:
C₇H₇Cl₂N₃O Molecular Weight: 220,056 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02187208 CAS-Nummer: 66999-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...