5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol, tech

Lieferant: Apollo Scientific

OR33018-1G
APOSOR33018-1GEA 1570 EUR
APOSOR33018-1G
5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol, tech
5-(Methoxymethyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-ol
Formel: C₉H₁₁N₃O₂
Molecular Weight: 193,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02661789
CAS-Nummer: 439111-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR