3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33019-1G
APOSOR33019-1GEA 1570 EUR
APOSOR33019-1G
3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one, tech
3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one
Formel: C₁₂H₁₂ClNO
Molecular Weight: 221,683 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00105289
CAS-Nummer: 478064-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR