N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide, tech

Lieferant: Apollo Scientific

OR33022-1G
APOSOR33022-1GEA 1570 EUR
APOSOR33022-1G
N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide, tech
N'-[(1E)-(Dimethylamino)methylidene]-1H,2H-naphtho[2,1-b]furan-2-carbohydrazide
Formel: C₁₆H₁₇N₃O₂
Molecular Weight: 283,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187225
CAS-Nummer: 478064-24-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR