5-[(Dimethylamino)methyl]-2,2-dimethyl-1,3-dioxane-4,6-dione, tech

Lieferant: Apollo Scientific

OR33023-1G
APOSOR33023-1GEA 1570 EUR
APOSOR33023-1G
5-[(Dimethylamino)methyl]-2,2-dimethyl-1,3-dioxane-4,6-dione, tech
5-[(Dimethylamino)methyl]-2,2-dimethyl-1,3-dioxane-4,6-dione
Formel: C₉H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02570817
CAS-Nummer: 227201-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR