Ethyl 2-(3-aminophenyl)indolizine-1-carboxylate, tech

Lieferant: Apollo Scientific

OR33024-1G
APOSOR33024-1GEA 1570 EUR
APOSOR33024-1G
Ethyl 2-(3-aminophenyl)indolizine-1-carboxylate, tech
Ethyl 2-(3-aminophenyl)indolizine-1-carboxylate
Formel: C₁₇H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02187336
CAS-Nummer: 478064-54-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR