2-Ethoxy-6-(furan-2-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33025-1G
APOSOR33025-1GEA 1570 EUR
APOSOR33025-1G
2-Ethoxy-6-(furan-2-yl)pyridine-3-carbonitrile, tech
2-Ethoxy-6-(furan-2-yl)pyridine-3-carbonitrile
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02570837
CAS-Nummer: 190579-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR