1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one, tech

Lieferant: Apollo Scientific

OR33026-1G
APOSOR33026-1GEA 1570 EUR
APOSOR33026-1G
1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one, tech
1-[6-(4-Methoxyphenoxy)-2-methylpyridin-3-yl]ethan-1-one
Formel: C₁₅H₁₅NO₃
Molecular Weight: 257,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02570895
CAS-Nummer: 478064-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR