1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one 95%

Lieferant: Apollo Scientific

OR33029-1G
APOSOR33029-1GEA 671 EUR
APOSOR33029-1G
1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one 95%
1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one
Formel: C₉H₉N₃O
Molecular Weight: 175,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00825993
CAS-Nummer: 64882-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR