5-Carboxy-2-chlorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-carboxyphenylboronic acid

OR3303-5G
APOSOR3303-5GEA 79.9 EUR
APOSOR3303-5G
5-Carboxy-2-chlorphenylboronsäure 98%
5-Carboxy-2-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200,3800 g/mol
Schmelzpunkt: 228…244 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689491
CAS-Nummer: 913835-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR