(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33030-1G
APOSOR33030-1GEA 1570 EUR
APOSOR33030-1G
(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₇H₂₀N₂O
Molecular Weight: 268,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012839
CAS-Nummer: 692287-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR