2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
Lieferant: Apollo Scientific
OR33031-1G
APOSOR33031-1GEA
1570
EUR
APOSOR33031-1G
2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol, tech
2-[3-(2H-1,3-Benzodioxol-5-yl)-1H-pyrazol-1-yl]ethan-1-ol
Formel:
C₁₂H₁₂N₂O₃ Molecular Weight: 232,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03617679 CAS-Nummer: 955976-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...