Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate 95%

Lieferant: Apollo Scientific

OR33032-1G
APOSOR33032-1GEA 671 EUR
APOSOR33032-1G
Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate 95%
Ethyl-1-(6-quinoxalinyl)-4-piperidinecarboxylate
Formel: C₁₆H₁₉N₃O₂
Molecular Weight: 285,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425801
CAS-Nummer: 453557-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR