2,3-Dihydro-1,4-benzoxathiin-6-ol 97%
Lieferant: Apollo Scientific
OR33034-1G
APOSOR33034-1GEA
1570
EUR
APOSOR33034-1G
2,3-Dihydro-1,4-benzoxathiin-6-ol 97%
2,3-Dihydro-1,4-benzoxathiin-6-ol
Formel:
C₈H₈O₂S Molecular Weight: 168,213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22571438 CAS-Nummer: 402911-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...