N-Benzyl-1H-indole-3-carboxamide 95%
Lieferant: Apollo Scientific
OR33035-1G
APOSOR33035-1GEA
1120
EUR
APOSOR33035-1G
N-Benzyl-1H-indole-3-carboxamide 95%
N-Benzyl-1H-indole-3-carboxamide
Formel:
C₁₆H₁₄N₂O Molecular Weight: 250,295 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03305733 CAS-Nummer: 61788-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...