6-(2,3-Dihydro-1H-inden-5-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
Lieferant: Apollo Scientific
OR33036-1G
APOSOR33036-1GEA
1570
EUR
APOSOR33036-1G
6-(2,3-Dihydro-1H-inden-5-yl)-2,3,4,5-tetrahydropyridazin-3-one, tech
6-(2,3-Dihydro-1H-inden-5-yl)-2,3,4,5-tetrahydropyridazin-3-one
Formel:
C₁₃H₁₄N₂O Molecular Weight: 214,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787574 CAS-Nummer: 866050-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...