5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33037-1G
APOSOR33037-1GEA 1570 EUR
APOSOR33037-1G
5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
5-(2,3-Dihydro-1H-inden-5-yl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₆H₁₂N₄
Molecular Weight: 260,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125854
CAS-Nummer: 866050-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR