4-(2,3-Dihydro-1H-inden-5-yl)pyrimidin-2-amine, tech

Lieferant: Apollo Scientific

OR33038-1G
APOSOR33038-1GEA 1570 EUR
APOSOR33038-1G
4-(2,3-Dihydro-1H-inden-5-yl)pyrimidin-2-amine, tech
4-(2,3-Dihydro-1H-inden-5-yl)pyrimidin-2-amine
Formel: C₁₃H₁₃N₃
Molecular Weight: 211,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787582
CAS-Nummer: 866050-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR