2-Ethoxy-5-methylbenzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ethoxy-5-methylbenzeneboronic acid, 2-Ethoxy-5-methylphenylboronic acid

OR3304-1G
APOSOR3304-1GEA 21.9 EUR
APOSOR3304-1G
2-Ethoxy-5-methylbenzeneboronic acid 95%
2-Ethoxy-5-methylbenzeneboronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
Schmelzpunkt: 98…101 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00196832
CAS-Nummer: 123291-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR