5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR33041-1G
APOSOR33041-1GEA 1570 EUR
APOSOR33041-1G
5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione, tech
5,5-Dimethyl-2-[(phenylamino)methylidene]cyclohexane-1,3-dione
Formel: C₁₅H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00512921
CAS-Nummer: 29974-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR