Methyl 2-(4-methoxyphenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech

Lieferant: Apollo Scientific

OR33042-1G
APOSOR33042-1GEA 1570 EUR
APOSOR33042-1G
Methyl 2-(4-methoxyphenyl)imidazo[1,2-a]pyridine-6-carboxylate, tech
Methyl 2-(4-methoxyphenyl)imidazo[1,2-a]pyridine-6-carboxylate
Formel: C₁₆H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04125892
CAS-Nummer: 866050-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR