Methyl 2-(1-methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoate, tech

Lieferant: Apollo Scientific

OR33048-1G
APOSOR33048-1GEA 1570 EUR
APOSOR33048-1G
Methyl 2-(1-methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoate, tech
Methyl 2-(1-methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoate
Formel: C₁₈H₁₅NO₃
Molecular Weight: 293,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256228
CAS-Nummer: 294184-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR