[(E)-2-(4-Chloro-2-nitrophenyl)ethenyl]dimethylamine, tech

Lieferant: Apollo Scientific

OR33050-1G
APOSOR33050-1GEA 1570 EUR
APOSOR33050-1G
[(E)-2-(4-Chloro-2-nitrophenyl)ethenyl]dimethylamine, tech
[(E)-2-(4-Chloro-2-nitrophenyl)ethenyl]dimethylamine
Formel: C₁₀H₁₁ClN₂O₂
Molecular Weight: 226,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05974962
CAS-Nummer: 32989-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR