2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR33052-1G
APOSOR33052-1GEA
538
EUR
APOSOR33052-1G
2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde 95%
2-Cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde
Formel:
C₁₂H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840949 CAS-Nummer: 1135282-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...