4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde 95%

Lieferant: Apollo Scientific

OR33056-1G OR33056-5G
APOSOR33056-1GEA 0 EUR
APOSOR33056-1G APOSOR33056-5G
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde 95%
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde
Formel: C₁₄H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07782410
CAS-Nummer: 342889-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR